Learn More
Accuflow Systems TUBING MFLEX STA-PURE #24 14
Catalog No. 501109110
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. L/S 24 Gore Sta-Pure Tubing - Series PCS for Masterflex Pumps, 14" per pack Custom extruded to fit Masterflex pumps Engineered for long life in peristaltic pump applications Lot-to-lot consistency provides
Catalog No. 50-110-9110 Supplier Accuflow Systems Supplier No. 9624124
Manufactured by Cole Parmer and sold by Accuflow. L/S 24 Gore Sta-Pure Tubing - Series PCS for Masterflex Pumps, 14" per pack Custom extruded to fit Masterflex pumps Engineered for long life in peristaltic pump applications Lot-to-lot consistency provides