Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Ismatec Pump Tubing Element, GORE STA-PURE 3.2 mm ID x 6.4 mm OD;
Catalog No. NC3246225
Encompass
Masterflex Ismatec Pump Tubing Element, GORE STA-PURE Series PFL, 3.2 mm ID x 6.4 mm OD; Dimensions: 3.2 mm ID x 6.4 mm OD; 355 mm L (13.9764 inches); Characteristics: High resilience and strength, low spallation and gas permeability. Opaque.
Catalog No. NC3246225 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9625208
Masterflex Ismatec Pump Tubing Element, GORE STA-PURE Series PFL, 3.2 mm ID x 6.4 mm OD; Dimensions: 3.2 mm ID x 6.4 mm OD; 355 mm L (13.9764 inches); Characteristics: High resilience and strength, low spallation and gas permeability. Opaque.