Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, PharmaPure, I/P 82; 25 Ft
Catalog No. NC1460478
Encompass
Masterflex PharmaPure Precision pump tubing has ultra-low spallation and low extractability to protect the purity of samples. This high-performance formulation is nontoxic and nonhemolytic making it ideal for sterile, filtration, and cell media applications. Low extractables and low gas permeability provides a protective barrier to chemicals that damage tubing while maintaining the integrity of your fluid.
Catalog No. NC1460478 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643582/DEL
Masterflex PharmaPure Precision pump tubing has ultra-low spallation and low extractability to protect the purity of samples. This high-performance formulation is nontoxic and nonhemolytic making it ideal for sterile, filtration, and cell media applications. Low extractables and low gas permeability provides a protective barrier to chemicals that damage tubing while maintaining the integrity of your fluid.