Learn More
AVANTOR MASTERFLEX (VWR LLC) ChemFluor TSPTFE-0125-031 PTFE Tubing, 1/16"" x 1/8""; 50 ft/Pk
Catalog No. NC9089429
Encompass
Applications: Chemical transfer and pharmaceutical processes. Characteristics: Most flexible fluoropolymer. Excellent chemical compatibility. Translucent. Certification: FDA compliant. Temperature range: ¿450 to 550¿F (¿268 to 288¿C). Sterilize: Autoclave, ethylene oxide, or dry heat.
Catalog No. NC9089429 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9600002
Applications: Chemical transfer and pharmaceutical processes. Characteristics: Most flexible fluoropolymer. Excellent chemical compatibility. Translucent. Certification: FDA compliant. Temperature range: ¿450 to 550¿F (¿268 to 288¿C). Sterilize: Autoclave, ethylene oxide, or dry heat.