Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Norprene, 1/8"" ID x 1/4"" OD; 50 ft
Catalog No. 501961037
Encompass
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275¿F (-60 to 135¿C). Can be sterilized with autoclave.
Catalog No. 50-196-1037 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641002
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275¿F (-60 to 135¿C). Can be sterilized with autoclave.