Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Norprene A-60-G, L/S 17; 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0640417/DEL
Encompass
Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance Characteristics: Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance, opaque black
Catalog No. NC9737329
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.