Learn More
Waters Corp Waters Tubing, Outlet, 0.004 in. ID, for Column Manager, with Fitting
Catalog No. 50466602 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Waters outlet tubing is designed for use with column managers. It features a 0.004 in. ID and is black in color, providing a reliable connection for your chromatographic system.
  • 0.004 in. ID column outlet tubing
  • Black in color
  • For column managers
  • Includes fitting
  • 1 unit per package
Catalog No. 50-466-602 Supplier Waters Corp Supplier No. 700004163
This Waters outlet tubing is designed for use with column managers. It features a 0.004 in. ID and is black in color, providing a reliable connection for your chromatographic system.
  • 0.004 in. ID column outlet tubing
  • Black in color
  • For column managers
  • Includes fitting
  • 1 unit per package