Learn More
Med Vet International Dynarex Oxygen Tubing, inline Water Trap, 10/Case
Catalog No. 502829189
Encompass_Preferred
Dynarex Resp-O2 In-Line Water Traps are designed to collect and trap accumulated moisture from oxygen tubing during respiratory/vent treatment and help prevent excess liquid from being inhaled. Traps and collects excess moisture from oxygen tubing Keeps liquid away from patients’ nasal passages Tapered fittings ensure a secure connection to the tubing Transparent body allows for clear monitoring of fluid Not made with natural rubber latex
Catalog No. 50-282-9189 Supplier Med Vet International Supplier No. 33302
Dynarex Resp-O2 In-Line Water Traps are designed to collect and trap accumulated moisture from oxygen tubing during respiratory/vent treatment and help prevent excess liquid from being inhaled. Traps and collects excess moisture from oxygen tubing Keeps liquid away from patients’ nasal passages Tapered fittings ensure a secure connection to the tubing Transparent body allows for clear monitoring of fluid Not made with natural rubber latex