Learn More
Scientific Commodities Inc TUBING PE 0.28X0.64 MM 100FT
Catalog No. NC9069194
Encompass_Preferred
Tubing, 0.28 I.D. x 0.64mm O.D.
Catalog No. NC9069194 Supplier Scientific Commodities Inc Supplier No. BB31695PE1
Tubing, 0.28 I.D. x 0.64mm O.D.