Learn More
Instech PE-20 tubing, .015x.043in (0.4x1.1mm), 30m (98ft) spool, non-sterile.
SDP
Catalog No. 50890048
Encompass
The Instech PBTPE‑20 is a PE‑20 low‑density polyethylene tubing with a .015x.043in (0.4x1.1mm) flow path used for research infusion and sampling applications. It provides excellent compound compatibility with low air permeability and moderate stiffness, and is supplied non‑sterile on a 30m (98ft) spool for general laboratory use.
Features
  • PE‑20 low‑density polyethylene tubing
  • .015x.043in (0.4x1.1mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Good chemical compatibility and low permeability
  • Stiff, lightweight construction for external connections
  • Not for intravascular or human use
Catalog No. 50-890-048 Supplier Instech Supplier No. BTPE20
 Restricted Item
Add to Cart
Add to Cart
The Instech PBTPE‑20 is a PE‑20 low‑density polyethylene tubing with a .015x.043in (0.4x1.1mm) flow path used for research infusion and sampling applications. It provides excellent compound compatibility with low air permeability and moderate stiffness, and is supplied non‑sterile on a 30m (98ft) spool for general laboratory use.
Features
  • PE‑20 low‑density polyethylene tubing
  • .015x.043in (0.4x1.1mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Good chemical compatibility and low permeability
  • Stiff, lightweight construction for external connections
  • Not for intravascular or human use