Learn More
Instech PE tubing, .018x.036in (0.5x0.9mm), 30m (98ft) spool, non-sterile. Fits 25ga.
SDP
Catalog No. 50890049
Encompass
The Instech BTPE‑25 is a PE‑25 low‑density polyethylene tubing sized at .018x.036in (0.5x0.9mm), designed to fit 25ga swivels, couplers, and other small‑diameter laboratory connectors. It provides excellent chemical compatibility with a stiff, low‑permeability flow path and is supplied non‑sterile on a 30m (98ft) spool for general external infusion or sampling applications.
Features
  • PE tubing sized for 25ga connections
  • .018x.036in (0.5x0.9mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Good chemical compatibility and low air permeability
  • Stiff, lightweight tubing for external use
  • Not for intravascular or human use
Catalog No. 50-890-049 Supplier Instech Supplier No. BTPE25
The Instech BTPE‑25 is a PE‑25 low‑density polyethylene tubing sized at .018x.036in (0.5x0.9mm), designed to fit 25ga swivels, couplers, and other small‑diameter laboratory connectors. It provides excellent chemical compatibility with a stiff, low‑permeability flow path and is supplied non‑sterile on a 30m (98ft) spool for general external infusion or sampling applications.
Features
  • PE tubing sized for 25ga connections
  • .018x.036in (0.5x0.9mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Good chemical compatibility and low air permeability
  • Stiff, lightweight tubing for external use
  • Not for intravascular or human use