Learn More
Instech PE-50 tubing, .023x.038in (0.6x1.0mm), 30m (98ft) spool, non-sterile. Fits 22ga.
SDP
Catalog No. 50890050
Encompass
The Instech BTPE‑50 is a PE‑50 low‑density polyethylene tubing with a .023x.038in (0.6x1.0mm) profile, providing a stiff, low‑permeability flow path ideal for external infusion or sampling connections. Designed to fit 22ga swivels, couplers, and luer‑based components, it is supplied non‑sterile on a 30m (98ft) spool for general laboratory research use.
Features
  • PE‑50 low‑density polyethylene tubing
  • .023x.038in (0.6x1.0mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Fits 22ga swivels, couplers, and connectors
  • Good compound compatibility and low air permeability
  • Not for intravascular or human use
Catalog No. 50-890-050 Supplier Instech Supplier No. BTPE50
The Instech BTPE‑50 is a PE‑50 low‑density polyethylene tubing with a .023x.038in (0.6x1.0mm) profile, providing a stiff, low‑permeability flow path ideal for external infusion or sampling connections. Designed to fit 22ga swivels, couplers, and luer‑based components, it is supplied non‑sterile on a 30m (98ft) spool for general laboratory research use.
Features
  • PE‑50 low‑density polyethylene tubing
  • .023x.038in (0.6x1.0mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Fits 22ga swivels, couplers, and connectors
  • Good compound compatibility and low air permeability
  • Not for intravascular or human use