Learn More
Instech PE-60 tubing, .030x.048in (0.8x1.2mm), 30m (98ft) spool, non-sterile. Fits 20ga.
SDP
Catalog No. 50890051
Encompass
The Instech BTPE‑60 is a PE‑60 low‑density polyethylene tubing with a .030x.048in (0.76x1.22mm) profile, providing a stiff, low‑permeability flow path ideal for external infusion or sampling applications. Designed to fit 20ga swivels, couplers, and luer‑based connectors, it is supplied non‑sterile on a 30m (98ft) spool for general laboratory research use.
Features
  • PE‑60 low‑density polyethylene tubing
  • .030x.048in (0.76x1.22mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Fits 20ga swivels, couplers, and connectors
  • Good compound compatibility and low air permeability
  • Not for intravascular or human use
Catalog No. 50-890-051 Supplier Instech Supplier No. BTPE60
The Instech BTPE‑60 is a PE‑60 low‑density polyethylene tubing with a .030x.048in (0.76x1.22mm) profile, providing a stiff, low‑permeability flow path ideal for external infusion or sampling applications. Designed to fit 20ga swivels, couplers, and luer‑based connectors, it is supplied non‑sterile on a 30m (98ft) spool for general laboratory research use.
Features
  • PE‑60 low‑density polyethylene tubing
  • .030x.048in (0.76x1.22mm) dimensions
  • 30m (98ft) non‑sterile spool
  • Fits 20ga swivels, couplers, and connectors
  • Good compound compatibility and low air permeability
  • Not for intravascular or human use