Learn More
Instech PE-90 tubing, .034x.050in (0.9x1.3mm), 30m (98ft) spool, non-sterile.
SDP
Catalog No. 50890052
Encompass
The Instech BTPE‑90 is a PE‑90 low‑density polyethylene tubing with a .034x.050in (0.9x1.3mm) profile, providing a stiff, low‑permeability flow path suitable for external infusion and sampling applications. Supplied non‑sterile on a 30m (98ft) spool, it offers excellent compound compatibility and is commonly used in laboratory research setups requiring larger‑bore PE tubing.
Features
  • PE‑90 low‑density polyethylene tubing
  • .034x.050in (0.9x1.3mm) dimensions
  • 30m (98ft) non‑sterile spool
  • High compound compatibility and low air permeability
  • Stiff, durable tubing for external use
  • Not for intravascular or human use
Catalog No. 50-890-052 Supplier Instech Supplier No. BTPE90
 Restricted Item
Add to Cart
Add to Cart
The Instech BTPE‑90 is a PE‑90 low‑density polyethylene tubing with a .034x.050in (0.9x1.3mm) profile, providing a stiff, low‑permeability flow path suitable for external infusion and sampling applications. Supplied non‑sterile on a 30m (98ft) spool, it offers excellent compound compatibility and is commonly used in laboratory research setups requiring larger‑bore PE tubing.
Features
  • PE‑90 low‑density polyethylene tubing
  • .034x.050in (0.9x1.3mm) dimensions
  • 30m (98ft) non‑sterile spool
  • High compound compatibility and low air permeability
  • Stiff, durable tubing for external use
  • Not for intravascular or human use