Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.007" ID, 1/16" OD, General Grade, Yellow. 25 Meter Roll. 1 EA.
Catalog No. 502271533
Encompass_Preferred
This General Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives and is often used in HPLC laboratories. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Yellow for 0.007" ( 0.18mm ) ID. Pressure rating: 5800 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1533 Supplier Microsolv Technology Corporation Supplier No. 4930507C
This General Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives and is often used in HPLC laboratories. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Yellow for 0.007" ( 0.18mm ) ID. Pressure rating: 5800 psi.