Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.020" ID, 1/16" OD, General Grade, Orange. 5 Meter Roll. 1 EA.
Catalog No. 502271535
Encompass_Preferred
This General Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives and is often used in HPLC laboratories. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Orange for 0.020" ( 0.50mm ) ID. Pressure rating: 4500 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1535 Supplier Microsolv Technology Corporation Supplier No. 4930520
This General Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives and is often used in HPLC laboratories. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Orange for 0.020" ( 0.50mm ) ID. Pressure rating: 4500 psi.