Learn More
Microsolv Technology Corporation Tubing Peek .005 Id 5M R Solid
Catalog No. 5001451637
Encompass_Preferred
Tubing, PEEK, 0.005 ID, 1/16 OD, General Grade, Red. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51637 Supplier Microsolv Technology Corporation Supplier No. 4930505
Tubing, PEEK, 0.005 ID, 1/16 OD, General Grade, Red. 5 Meter Roll. MicroSolv Brand. 1/EA.