Learn More
Microsolv Technology Corporation Tubing Peek .005 Id 5M R Strpd
Catalog No. 5001451524
Encompass_Preferred
Tubing, PEEK, 0.005 ID, 1/16 OD, HPLC Grade, Red Stripe. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51524 Supplier Microsolv Technology Corporation Supplier No. 4730505
Tubing, PEEK, 0.005 ID, 1/16 OD, HPLC Grade, Red Stripe. 5 Meter Roll. MicroSolv Brand. 1/EA.