Learn More
Microsolv Technology Corporation Tubing Peek .007 Id 5M Y Solid
Catalog No. 5001451639
Encompass_Preferred
Tubing, PEEK, 0.007 ID, 1/16 OD, General Grade, Yellow. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51639 Supplier Microsolv Technology Corporation Supplier No. 4930507
Tubing, PEEK, 0.007 ID, 1/16 OD, General Grade, Yellow. 5 Meter Roll. MicroSolv Brand. 1/EA.