Learn More
Microsolv Technology Corporation Tubing Peek.010 Id 25M B Solid
Catalog No. 5001451641
Encompass_Preferred
Tubing, PEEK, 0.010 ID, 1/16 OD, General Grade, Blue. 25 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51641 Supplier Microsolv Technology Corporation Supplier No. 4930510C
Tubing, PEEK, 0.010 ID, 1/16 OD, General Grade, Blue. 25 Meter Roll. MicroSolv Brand. 1/EA.