Learn More
Microsolv Technology Corporation Tubing Peek.020 Id 25M O Solid
Catalog No. 5001451643
Encompass_Preferred
Tubing, PEEK, 0.020 ID, 1/16 OD, General Grade, Orange. 25 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51643 Supplier Microsolv Technology Corporation Supplier No. 4930520C
Tubing, PEEK, 0.020 ID, 1/16 OD, General Grade, Orange. 25 Meter Roll. MicroSolv Brand. 1/EA.