Learn More
Microsolv Technology Corporation Tubing Peek .020 Id 5M O Strpd
Catalog No. 5001451530
Encompass_Preferred
Tubing, PEEK, 0.020 ID, 1/16 OD, HPLC Grade, Orange Stripe. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51530 Supplier Microsolv Technology Corporation Supplier No. 4730520
Tubing, PEEK, 0.020 ID, 1/16 OD, HPLC Grade, Orange Stripe. 5 Meter Roll. MicroSolv Brand. 1/EA.