Learn More
Microsolv Technology Corporation Tubing Peek .030 Id 5M G Strpd
Catalog No. 5001451532
Encompass_Preferred
Tubing, PEEK, 0.030 ID, 1/16 OD, HPLC Grade, Green Stripe. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51532 Supplier Microsolv Technology Corporation Supplier No. 4730530
Tubing, PEEK, 0.030 ID, 1/16 OD, HPLC Grade, Green Stripe. 5 Meter Roll. MicroSolv Brand. 1/EA.