Learn More
Trajan Scientific VICI Tubing, PEEK, 1/16" OD x 0.005", Red, UOM 25FT
Catalog No. NC1264339
Encompass_Preferred
VICI Valco Tubing, PEEK, 1/16x0.005", Red, UOM 25FTPEEK Tubing has the strength Required to Withstand continuous use at HPLC Pressure Without swelling or bursting, and is not affected by halide salts, High strength buffers, or other aggressive mobile phases that corrode Stainless Steel. The polymer surface will not leach metal ions into the eluent or extract metal-sensitive Components from the Sample. Note however that dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentRated nitric and sµlphuric acid will attack PEEK.Tolerance is ±.0005" For IDs up to .010", and ±.001" For larger IDs. Made in the USA.
Catalog No. NC1264339 Supplier Trajan Scientific Supplier No. 100.JRT600725
VICI Valco Tubing, PEEK, 1/16x0.005", Red, UOM 25FTPEEK Tubing has the strength Required to Withstand continuous use at HPLC Pressure Without swelling or bursting, and is not affected by halide salts, High strength buffers, or other aggressive mobile phases that corrode Stainless Steel. The polymer surface will not leach metal ions into the eluent or extract metal-sensitive Components from the Sample. Note however that dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentRated nitric and sµlphuric acid will attack PEEK.Tolerance is ±.0005" For IDs up to .010", and ±.001" For larger IDs. Made in the USA.