Learn More
Microsolv Technology Corporation Tubing Peek 1/16X005 Supert
Catalog No. 5001451540
Encompass_Preferred
Tubing, PEEK, 0.005 ID, 1/16 OD, Super-T Grade, Red Stripes. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51540 Supplier Microsolv Technology Corporation Supplier No. 47305ST05
Tubing, PEEK, 0.005 ID, 1/16 OD, Super-T Grade, Red Stripes. 5 Meter Roll. MicroSolv Brand. 1/EA.