Learn More
Microsolv Technology Corporation Tubing Peek 1/8Od X 1/16 Id
Catalog No. 5001451546
Encompass_Preferred
Tubing, PEEK, 1/16 ID, 1/8 OD, Natural Color. 10 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51546 Supplier Microsolv Technology Corporation Supplier No. 47310100
Tubing, PEEK, 1/16 ID, 1/8 OD, Natural Color. 10 Meter Roll. MicroSolv Brand. 1/EA.