Learn More
IDEX Health & Science Upchurch TUBING PEEK 100' ROLL OF 1533
Catalog No. NC1919803
Encompass_Preferred
Idex PEEK Chromatography Tubing, Green, Diameter Inner: 0.03 in., Overall Length: 100 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919803 Supplier IDEX Health & Science Upchurch Supplier No. 1533XL
Idex PEEK Chromatography Tubing, Green, Diameter Inner: 0.03 in., Overall Length: 100 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications