Learn More
IDEX Health & Science Upchurch TUBING PEEK 100' ROLL OF 1535
Catalog No. NC1919816
Encompass_Preferred
Idex PEEK Chromatography Tubing, Natural, Diameter Inner: 0.005 in., Overall Length: 100 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919816 Supplier IDEX Health & Science Upchurch Supplier No. 1535XL
Idex PEEK Chromatography Tubing, Natural, Diameter Inner: 0.005 in., Overall Length: 100 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications