Learn More
DSI LLC Tubing, PEEK, 1/16'' x 0.13 mm ID, solid red
Catalog No. 502255511
Encompass_Preferred
Tubing, PEEK, 1/16'' x 0.13 mm ID, solid red
Catalog No. 50-225-5511 Supplier DSI LLC Supplier No. JRT6007
Tubing, PEEK, 1/16'' x 0.13 mm ID, solid red