Learn More
DSI LLC Tubing, PEEK, 1/32" x 75 um / .003" ID, green striped, 3 m/pkg
Catalog No. 502255410
Encompass_Preferred
Tubing, PEEK, 1/32" x 75 um / .003" ID, green striped, 3 m/pkg
Catalog No. 50-225-5410 Supplier DSI LLC Supplier No. JRT5703M3
Tubing, PEEK, 1/32" x 75 um / .003" ID, green striped, 3 m/pkg