Learn More
IDEX Health & Science Upchurch TUBING PEEK 50' ROLL OF 1531
Catalog No. NC1919753
Encompass_Preferred
Idex PEEK Chromatography Tubing, Natural, Diameter Inner: 0.01 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919753 Supplier IDEX Health & Science Upchurch Supplier No. 1531L
Idex PEEK Chromatography Tubing, Natural, Diameter Inner: 0.01 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications