Learn More
IDEX Health & Science Upchurch TUBING PEEK 50' ROLL OF 1531B
Catalog No. NC1919776
Encompass_Preferred
Idex PEEK Chromatography Tubing, Blue, Diameter Inner: 0.01 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919776 Supplier IDEX Health & Science Upchurch Supplier No. 1531BL
Idex PEEK Chromatography Tubing, Blue, Diameter Inner: 0.01 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications