Learn More
IDEX Health & Science Upchurch TUBING PEEK 50' ROLL OF 1536
Catalog No. NC1919821
Encompass_Preferred
Idex PEEK Chromatography Tubing, Yellow, Diameter Inner: 0.007 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919821 Supplier IDEX Health & Science Upchurch Supplier No. 1536L
Idex PEEK Chromatography Tubing, Yellow, Diameter Inner: 0.007 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications