Learn More
IDEX Health & Science Upchurch TUBING PEEK 50FT ROLL OF 1535
Catalog No. NC1919812
Encompass_Preferred
Idex PEEK Chromatography Tubing, Red, Diameter Inner: 0.005 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919812 Supplier IDEX Health & Science Upchurch Supplier No. 1535L
Idex PEEK Chromatography Tubing, Red, Diameter Inner: 0.005 in., Overall Length: 50 ft., Pressure: 7000 psi, PEEK Tubing, For Use With: For High-Pressure Applications