Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.75mm ID, 1/8" OD, Natural Color. 10 Meter Roll. 1 EA.
Catalog No. 502271478
Encompass_Preferred
This wide outer diameter, Laboratory Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color: Natural. Pressure rating: 5000 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1478 Supplier Microsolv Technology Corporation Supplier No. 4731075
This wide outer diameter, Laboratory Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color: Natural. Pressure rating: 5000 psi.