Learn More
Trajan Scientific Tubing PEEK Black Medical 1/EA
SDP
Supplier:  Trajan Scientific 1301004006100F
Encompass_Preferred
1/32 OD x 0 004 ID x 100 length PEEK tubing black
Catalog No. 50-004-94163