Learn More
Trajan Scientific Tubing PEEK Black Medical 1/EA
SDP
Catalog No. 5000494585
Encompass_Preferred
360 m OD x 75 m (0 003 ) ID x 10 length PEEK tubing black
Catalog No. 50-004-94585 Supplier Trajan Scientific Supplier No. 130400300610F
360 m OD x 75 m (0 003 ) ID x 10 length PEEK tubing black