Learn More
Trajan Scientific Tubing PEEK Black Medical 1/EA
SDP
Catalog No. 5000494366
Encompass_Preferred
360 m OD x 75 m (0 003 ) ID x 100 length PEEK tubing black
Catalog No. 50-004-94366 Supplier Trajan Scientific Supplier No. 1304003006100F
360 m OD x 75 m (0 003 ) ID x 100 length PEEK tubing black