Learn More
Trajan Scientific Tubing PEEK Blue Medical 1/EA
SDP
Catalog No. 5000494370
Encompass_Preferred
1/16 OD x 0 010 ID x 150 m length PEEK tubing blue
Catalog No. 50-004-94370 Supplier Trajan Scientific Supplier No. 1302010003150M
1/16 OD x 0 010 ID x 150 m length PEEK tubing blue