Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.010" ID, 1/16" OD, Super-T Grade, Blue Stripes. 5 Meter Roll. 1 EA.
Catalog No. 502271473
Encompass_Preferred
This Super-T™ grade ( Premium ), PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The broken yellow stripes are added only to the outside of the tubing, leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.025mm. Color Code: Blue, Stripes for 0.010" ( 0.25mm ) ID. Pressure rating: 5600 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1473 Supplier Microsolv Technology Corporation Supplier No. 47305ST10
This Super-T™ grade ( Premium ), PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The broken yellow stripes are added only to the outside of the tubing, leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.025mm. Color Code: Blue, Stripes for 0.010" ( 0.25mm ) ID. Pressure rating: 5600 psi.