Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.010" ID, 1/16" OD, HPLC Grade, Blue Stripe. 25 Meter Roll. 1 EA.
Catalog No. 502271460
Encompass_Preferred
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The red stripe is added only to the outside of the tubing leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Blue Stripe for 0.010" ( 0.25mm ) ID. Pressure rating: 5600 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1460 Supplier Microsolv Technology Corporation Supplier No. 4730510C
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The red stripe is added only to the outside of the tubing leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Blue Stripe for 0.010" ( 0.25mm ) ID. Pressure rating: 5600 psi.