Learn More
Microsolv Technology Corporation Tubing, PEEK, Coiled, 0.007" ID, 1/16" OD. 1585mm L, General Grade, Yellow, with Tubing Connectors. 1 EA.
Catalog No. 502271552
Encompass_Preferred
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 50mm wide when resting and 300mm when fully extended, with a total tubing length of 1585mm, end to end. Color Coded: Yellow for 0.007" (0.18mm) ID
This item is not returnable. View return policy
Catalog No. 50-227-1552 Supplier Microsolv Technology Corporation Supplier No. 49308503Y
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 50mm wide when resting and 300mm when fully extended, with a total tubing length of 1585mm, end to end. Color Coded: Yellow for 0.007" (0.18mm) ID