Learn More
Microsolv Technology Corporation Tubing, PEEK, Coiled, 0.030" ID, 1/16" OD. 945mm L, General Grade, Green, with Tubing Connectors. 1 EA.
Catalog No. 502271545
Encompass_Preferred
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 20mm wide when resting and 120mm when fully extended, with a total tubing length of 945mm, end to end. Color Coded: Green for 0.030" (0.75mm) ID
This item is not returnable. View return policy
Catalog No. 50-227-1545 Supplier Microsolv Technology Corporation Supplier No. 49306504G
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 20mm wide when resting and 120mm when fully extended, with a total tubing length of 945mm, end to end. Color Coded: Green for 0.030" (0.75mm) ID