Learn More
Microsolv Technology Corporation Tubing, PEEK, Coiled, 0.030" ID, 1/16" OD. 710mm L, General Grade, Green, with Tubing Connectors. 1 EA.
Catalog No. 502271541
Encompass_Preferred
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 10mm wide when resting and 30mm when fully extended, with a total tubing length of 700mm, end to end. Color Coded: Green for 0.030" (0.75mm) ID
This item is not returnable. View return policy
Catalog No. 50-227-1541 Supplier Microsolv Technology Corporation Supplier No. 49305504G
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 10mm wide when resting and 30mm when fully extended, with a total tubing length of 700mm, end to end. Color Coded: Green for 0.030" (0.75mm) ID