Learn More
Trajan Scientific Tubing PEEK Green Medical 1/EA
SDP
Catalog No. 5000494574
Encompass_Preferred
360 m OD x 125 m (0 005 ) ID x 10 length PEEK tubing green
Catalog No. 50-004-94574 Supplier Trajan Scientific Supplier No. 130400500510F
360 m OD x 125 m (0 005 ) ID x 10 length PEEK tubing green