Learn More
Trajan Scientific Tubing PEEK Green Medical 1/EA
SDP
Catalog No. 5000494371
Encompass_Preferred
1/16 OD x 0 030 ID x 100 m length PEEK tubing green
Catalog No. 50-004-94371 Supplier Trajan Scientific Supplier No. 1302030005100M
1/16 OD x 0 030 ID x 100 m length PEEK tubing green