Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.040" ID, 1/16" OD, HPLC Grade, Grey Stripe. 25 Meter Roll. 1 EA.
Catalog No. 502271464
Encompass_Preferred
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The red stripe is added only to the outside of the tubing leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Grey Stripe for 0.040" ( 1.0mm ) ID. Pressure rating: 2400 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1464 Supplier Microsolv Technology Corporation Supplier No. 4730540C
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The red stripe is added only to the outside of the tubing leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Grey Stripe for 0.040" ( 1.0mm ) ID. Pressure rating: 2400 psi.