Learn More
Trajan Scientific Tubing PEEK Natural Medical 1/EA
SDP
Catalog No. 5000494575
Encompass_Preferred
360 m OD x 50 m (0 002 ) ID x 100 length PEEK tubing natural
Catalog No. 50-004-94575 Supplier Trajan Scientific Supplier No. 1304002000100F
360 m OD x 50 m (0 002 ) ID x 100 length PEEK tubing natural