Learn More
Trajan Scientific Tubing PEEK Natural Medical 1/EA
SDP
Supplier:  Trajan Scientific 130100250010F
Encompass_Preferred
1/32 OD x 0 0025 ID x 10 length PEEK tubing natural
Catalog No. 50-004-93980