Learn More
Trajan Scientific Tubing PEEK Natural Medical 1/EA
SDP
Supplier:  Trajan Scientific 130201500050F
Encompass_Preferred
1/16 OD x 0 015 ID x 50 length PEEK tubing natural
Catalog No. 50-004-94562